
2,6-Di-tert-butyl-4-methoxyphenol ≥98.0% (by GC) oxidation inhibitor
Catalog # TCIAD1811-25G
Supplier: TCI
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:25 g
- Seq:3308
Specifications
Specification Test Results
- Appearance:White to Light yellow to Light orange powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:101.0 to 108.0 deg-C
Specification Test Results
About this item
:
:236.35 g/mol
Boiling Pt:140°C
Melting Pt:105.4 °C
EINECS:207-693-0
:GHS07
CAS Number:489-01-0
Formula:C₁₅H₂₄O₂
MDL Number:MFCD00008824