Order Entry
Italy
ContactUsLinkComponent
 
 
α,α,α,3-Tetrafluoro-p-toluic acid ≥98.0% (by GC, titration analysis)
 :  TCI
 
undefined
 
α,α,α,3-Tetrafluoro-p-toluic acid ≥98.0% (by GC, titration analysis)
 :  TCI

 

  • Appearance:
    White to Light yellow powder to crystal
  • Purity (GC):
    min. 98.0 %
  • Purity (Neutralization titration):
    min. 98.0 %
  • Melting point:
    174.0 to 178.0 deg-C

 

Frequently Bought Together