Order Entry
Italy
ContactUsLinkComponent
1,4-Dimethoxy-2-fluorobenzene ≥97.0% (by GC)
Irritant
1,4-Dimethoxy-2-fluorobenzene ≥97.0% (by GC)
Catalog # TCIAF0633-5G
Supplier:  TCI
CAS Number:  
undefined
Irritant
1,4-Dimethoxy-2-fluorobenzene ≥97.0% (by GC)
Catalog # TCIAF0633-5G
Supplier:  TCI
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

Specification Test Results

  • Appearance:
    White to Almost white powder to lump
  • Purity (GC):
    min. 97.0 %

Specification Test Results

About this item

 :
 :156.16 g/mol
Boiling Pt:121°C
Melting Pt:23…26 °C
 :GHS07
CAS Number:82830-49-7
Formula:C₈H₉FO₂
MDL Number:MFCD00042487