
N-(1-Naphthyl)ethylenediamine dihydrochloride ≥98.0% (by HPLC, total nitrogen) for biochemical research
Catalog # TCIAN0869-5G
Supplier: TCI
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:5 g
- Seq:3173
Specifications
Specification Test Results
- Appearance:White to Light yellow to Light orange powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (with Total Nitrogen):min. 98.0 % (calcd.on anh.substance)
- Water:max. 7.0 %
- Nitrite ion detection test (Griess method):to pass test
Specification Test Results
About this item
:
:259.18 g/mol
Merck Index:14,06409
Melting Pt:190°C
EINECS:215-981-2
:GHS07
Storage Temperature:Ambient
CAS Number:1465-25-4
Formula:C₁₂H₁₆Cl₂N₂
MDL Number:MFCD00012556