Order Entry
Italy
ContactUsLinkComponent
 
 
 
1-Chloro-2-butene (mixture of cis and trans isomers) >62.0% (by GC) (contains 38% 3-Chloro-1-butene at maximum)
 :  TCI
 
undefined
 
 
1-Chloro-2-butene (mixture of cis and trans isomers) >62.0% (by GC) (contains 38% 3-Chloro-1-butene at maximum)
 :  TCI

 

  • Appearance:
    Colorless to Light yellow clear liquid
  • Purity (GC):
    min. 62.0% (total of isomers)
  • 3-Chloro-1-butene:
    max. 38.0 %

 

Frequently Bought Together