Order Entry
Italy
ContactUsLinkComponent
Glutaraldehyde ∼50% ∼5.6 mol/L in water
Corrosive
Toxic
Health
Environment
Glutaraldehyde ∼50% ∼5.6 mol/L in water
Supplier:  TCI
Certificates
undefined
Corrosive
Toxic
Health
Environment
Glutaraldehyde ∼50% ∼5.6 mol/L in water
Supplier:  TCI

Specification Test Results

  • Appearance:
    Colorless to Light yellow clear liquid
  • Concentration (Oxime Formation):
    48.0 to 52.0 w/w%
  • pH:
    3.0 to 6.0 (25 deg-C)
  • Acid Value:
    max. 3.0

Specification Test Results

Related Information

Frequently Bought Together