Order Entry
Italy
ContactUsLinkComponent
1,2-Cyclododecanediol cis- and trans-mixture ≥87.0% (by GC)
1,2-Cyclododecanediol cis- and trans-mixture ≥87.0% (by GC)
Catalog # TCIAC1497-1G
Supplier:  TCI
CAS Number:  
undefined
1,2-Cyclododecanediol cis- and trans-mixture ≥87.0% (by GC)
Catalog # TCIAC1497-1G
Supplier:  TCI
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

Specification Test Results

  • Appearance:
    White to Almost white powder to crystal
  • Purity (GC):
    min. 87.0% (cis-, trans- mixture)
  • Melting point:
    121.0 to 125.0 deg-C

Specification Test Results

About this item

 :
 :200.32 g/mol
EINECS:239-256-5
CAS Number:15199-41-4
Formula:C₁₂H₂₄O₂
MDL Number:MFCD01321150