
Phthalic acid ≥99.5%
N985-500G
:
- Pack type:Plastic bottle
- Pk:500 g
- Seq:3645
- Assay:99.5%
- Chloride:= 0.001%
- Heavy Metals (as Pb):= 0.001%
- Insoluble Matter:= 0.05%
- Iron:= 0.001%
- Nitrate:= 0.005%
- Residue after Ignition:= 0.02%
- Sulphate:= 0.005%
- Water:= 0.5%
:
:166.13 g/mol
:12,07527
:315…317 °C (1013 hPa)
:79 °C
:1.576 g/cm³ (20 °C)
:Ambient
:201-873-2
:GHS07
:88-99-3
:C₈H₆O₄
:MFCD00002467