

Di-tert-butyl pyrocarbonate ≥95.0% (by titrimetric analysis) BOC-reagent for amino acid
Supplier: TCI
Certificates
About this item
CAS Number:24424-99-5
:
Formula:C₁₀H₁₈O₅
MDL Number:MFCD00008805
:218.25 g/mol
Boiling Pt:235 °C (1013 hPa)
Melting Pt:20…23 °C
Density:1,02 g/cm³ (20 °C)
Flash Pt:47°C
Storage Temperature:Refrigerator
EINECS:246-240-1
UN:2930
ADR:6.1,II
:GHS02
:GHS06
Specification Test Results
- Appearance:White or Colorless to Light yellow powder to lump to clear liquid
- Purity (Morpholine Method):min. 95.0 %
Specification Test Results
Related Information
Frequently Bought Together