

1,1'-Carbonyldiimidazole ≥97.0% (by titrimetric analysis) coupling agent for peptides synthesis
: TCI
:530-62-1
:
:C₇H₆N₄O
:MFCD00005286
:162.15 g/mol
:14,01819
:111…116 °C
:Refrigerator
:208-488-9
:1759
:8,III
:GHS05
:GHS07
- Environmentally Preferable:Safer Human and Environmental Health
- Appearance:White to Light yellow powder to crystal
- Purity (Neutralization titration):min. 97.0 %
- Melting point:114.0 to 121.0 deg-C
Frequently Bought Together