Order Entry
Italy
ContactUsLinkComponent
Pentadecan-15-olide ≥98.0% (by GC)
Pentadecan-15-olide ≥98.0% (by GC)
Catalog # TCIAP0985-25G
Supplier:  TCI
CAS Number:  
undefined
Pentadecan-15-olide ≥98.0% (by GC)
Catalog # TCIAP0985-25G
Supplier:  TCI
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    25 g
  • Seq:
    3308

Specifications

Specification Test Results

  • Appearance:
    White or Colorless to Light yellow powder to lump to clear liquid
  • Purity (GC):
    min. 98.0 %

Specification Test Results

About this item

 :
 :240.39 g/mol
Merck Index:14,03910
EINECS:203-354-6
CAS Number:106-02-5
Formula:C₁₅H₂₈O₂
MDL Number:MFCD00039667