
2,3-Dichloro-5,6-dicyano-p-benzoquinone ≥97.0% (by titrimetric analysis)
Supplier: TCI
Certificates
About this item
CAS Number:84-58-2
:
Formula:C₈Cl₂N₂O₂
MDL Number:MFCD00001593
:227.01 g/mol
Merck Index:14,03063
Boiling Pt:396 °C (1013 hPa)
Melting Pt:209…214 °C
Density:1,713 g/cm³ (20 °C)
Storage Temperature:Refrigerator
EINECS:201-542-2
UN:3439
ADR:6.1,III
:GHS06
Specification Test Results
- Appearance:Yellow to Amber to Dark purple powder to crystal
- Purity (Iodometric Titration):min. 97.0 %
Specification Test Results
Related Information
Frequently Bought Together