
2-Bromophenylboronic acid (contains varying amounts of Anhydride)
Catalog # TCIAB2889-5G
Supplier: TCI
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:5 g
- Seq:3173
Specifications
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (Neutralization titration):97.0 to 110.0 %
Specification Test Results
About this item
:
:200.83 g/mol
:GHS07
CAS Number:244205-40-1
Formula:C₆H₆BBrO₂
MDL Number:MFCD01114672