2,6-Dibromo-9,10-anthraquinone ≥95.0% (by HPLC)
Supplier: TCI
Certificates
About this item
CAS Number:633-70-5
:
Formula:C₁₄H₆Br₂O₂
MDL Number:MFCD08276344
:366.0042 g/mol
Melting Pt:290°C
Specification Test Results
- Appearance:Light yellow to Brown powder to crystal
- Purity (HPLC):min. 95.0 area%
Specification Test Results
Frequently Bought Together