
2,6-Dichlorobenzyl methyl ether
Catalog # TCIAD2109-25G
Supplier: TCI
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:25 g
- Seq:3308
Specifications
Specification Test Results
- Appearance:Colorless to Almost colorless clear liquid
- Purity (GC):min. 97.0 %
Specification Test Results
About this item
:
Boiling Pt:125°C
EINECS:251-541-6
:GHS07
CAS Number:33486-90-7
Formula:C₈H₈Cl₂O
MDL Number:MFCD00000578