Methyl-3,3-dimethoxypropionate ≥98.0% (by GC)
Catalog # TCIAD1858-25G
Supplier: TCI
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:25 g
- Seq:3308
Specifications
Specification Test Results
- Purity (GC):min. 98.0 %
- Appearance:Colorless to Almost colorless clear liquid
Specification Test Results
About this item
:
:148.16 g/mol
Boiling Pt:80°C
Flash Pt:67°C
EINECS:231-055-0
CAS Number:7424-91-1
Formula:C₆H₁₂O₄
MDL Number:MFCD00010650