Order Entry
Italy
ContactUsLinkComponent
Methyl-3,3-dimethoxypropionate 99%
Irritant
Methyl-3,3-dimethoxypropionate 99%
Certificates
undefined
Irritant
Methyl-3,3-dimethoxypropionate 99%

Specification Test Results

  • Appearance:
    Clear colorless Liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.4090 to 1.4110 (20°C, 589 nm)
  • GC:
    ≥98.5 %

Specification Test Results

Related Information

Frequently Bought Together