Order Entry
Italy
ContactUsLinkComponent
Tetrabutylammonium hydroxide 37% in methanol
Tetrabutylammonium hydroxide 37% in methanol
Supplier:  TCI
Certificates
undefined
Tetrabutylammonium hydroxide 37% in methanol
Supplier:  TCI

Specification Test Results

  • Appearance:
    Colorless to Light yellow to Light orange clear liquid
  • Concentration (Neutralization titration):
    37.0 to 40.7 w/w%

Specification Test Results

Frequently Bought Together