



Tetrabutylammonium hydroxide 10% in isopropanol (IPA)
Supplier: TCI
Certificates
About this item
CAS Number:2052-49-5
:
Formula:C₁₆H₃₇NO
MDL Number:MFCD00009425
:259.48 g/mol
Storage Temperature:Refrigerator
EINECS:218-147-6
UN:2924
ADR:3,II
:GHS02
:GHS05
:GHS07
:GHS08
Specification Test Results
- Appearance:Colorless to Light orange to Yellow clear liquid to slightly cloudy liquid
- Concentration (Neutralization titration):10.0 to 11.0 w/w%
Specification Test Results
Frequently Bought Together