Order Entry
Italy
ContactUsLinkComponent
 
Tetrabutylammonium hydroxide 10% in water
Catalog # TCIAT0216-25G
Supplier:  TCI
CAS Number:  
undefined
Tetrabutylammonium hydroxide 10% in water
Catalog # TCIAT0216-25G
Supplier:  TCI
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    25 g
  • Seq:
    3308

Specifications

Specification Test Results

  • Appearance:
    Colorless to Almost colorless clear liquid
  • Concentration (Neutralization titration):
    10.0 to 11.0 w/w%
  • Bromide:
    max. 0.005 %

Specification Test Results

About this item

 :
 :259.48 g/mol
Storage Temperature:Refrigerator
EINECS:218-147-6
UN:3267
ADR:8,II
 :GHS05
CAS Number:2052-49-5
Formula:C₁₆H₃₇NO
MDL Number:MFCD00009425