
3,6-Dichloropicolinic acid ≥98.0% (by GC, titration analysis)
Catalog # TCIAD4117-1G
Supplier: TCI
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 g
- Seq:3047
Specifications
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 98.0 %
- Purity (Neutralization titration):min. 98.0 %
- Melting point:149.0 to 153.0 deg-C
Specification Test Results
About this item
:
:192.00 g/mol
Merck Index:14,02400
EINECS:216-935-4
:GHS05
CAS Number:1702-17-6
Formula:C₆H₃Cl₂NO₂
MDL Number:MFCD00078655