

Di-n-octyl phthalate ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:117-84-0
:
Formula:C₂₄H₃₈O₄
MDL Number:MFCD00015292
:390.56 g/mol
Boiling Pt:380°C
Flash Pt:219°C
EINECS:204-214-7
:GHS07
:GHS08
Specification Test Results
- Appearance:Colorless to Light yellow to Light orange clear liquid
- Purity (GC):min. 98.0 %
Specification Test Results
Frequently Bought Together