Order Entry
Italy
ContactUsLinkComponent
Di-n-octyl phthalate
Di-n-octyl phthalate
Catalog # PROOCILULM61291.2
CAS Number:  
Di-n-octyl phthalate
Catalog # PROOCILULM61291.2
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1,2 mL
  • Seq:
    2080

Specifications

About this item

 :
 :390.56 g/mol
EINECS:204-214-7
CAS Number:117-84-0
Formula:C₂₄H₃₈O₄
MDL Number:MFCD00015292