4-Chloropyridine-N-oxide ≥97.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:1121-76-2
:
Formula:C₅H₄ClNO
MDL Number:MFCD00047425
EINECS:214-336-2
:GHS07
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 97.0 %
Specification Test Results
Frequently Bought Together