Order Entry
Italy
ContactUsLinkComponent
4,7-Dibromo-3-hydroxy-2-naphthoic acid ≥98.0% (by GC, titration analysis)
Irritant
4,7-Dibromo-3-hydroxy-2-naphthoic acid ≥98.0% (by GC, titration analysis)
Catalog # TCIAD3646-5G
Supplier:  TCI
CAS Number:  
undefined
Irritant
4,7-Dibromo-3-hydroxy-2-naphthoic acid ≥98.0% (by GC, titration analysis)
Catalog # TCIAD3646-5G
Supplier:  TCI
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

Specification Test Results

  • Appearance:
    Light yellow to Yellow to Green powder to crystal
  • Purity (GC):
    min. 98.0 %
  • Purity (Neutralization titration):
    min. 98.0 %

Specification Test Results

About this item

 :
 :345.98 g/mol
Melting Pt:253°C
EINECS:217-214-7
 :GHS07
CAS Number:1779-10-8
Formula:C₁₁H₆Br₂O₃
MDL Number:MFCD00046367