Order Entry
Italy
ContactUsLinkComponent
Benzaldehyde azine ≥98.0% (by GC, titration analysis)
Benzaldehyde azine ≥98.0% (by GC, titration analysis)
Catalog # TCIAB0006-25G
Supplier:  TCI
CAS Number:  
undefined
Benzaldehyde azine ≥98.0% (by GC, titration analysis)
Catalog # TCIAB0006-25G
Supplier:  TCI
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    25 g
  • Seq:
    3308

Specifications

Specification Test Results

  • Appearance:
    White to Yellow powder to crystal
  • Purity (GC):
    min. 98.0 %
  • Purity (Nonaqueous Titration):
    min. 98.0 %
  • Melting point:
    93.0 to 96.0 deg-C
  • Solubility in hot Methanol:
    almost transparency

Specification Test Results

About this item

 :
 :208.27 g/mol
Storage Temperature:Refrigerator
EINECS:209-627-6
CAS Number:588-68-1
Formula:C₁₄H₁₂N₂
MDL Number:MFCD00016876