Order Entry
Italy
ContactUsLinkComponent
4,4'-Anisoin for synthesis
4,4'-Anisoin for synthesis
Catalog # 8.41595.0025
Supplier:  Merck
CAS Number:  
4,4'-Anisoin for synthesis
Catalog # 8.41595.0025
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    25 g
  • Seq:
    3308

Specifications

Specification Test Results

  • Melting Point:
    111 - 112 °C
  • Molar Mass:
    272.3 g/mol
  • Synonyms:
    4,4'-Dimethoxybenzoin
  • Hill Formula:
    C16H16O4

Specification Test Results

About this item

 :
 :272.30 g/mol
 :M-Clarity™ quality level = MQ200
EINECS:204-330-8
CAS Number:119-52-8
Formula:C₁₆H₁₆O₄
MDL Number:MFCD00008411