
4,5-Dimethoxy-2-fluorobenzonitrile ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:119396-88-2
:
Formula:C₉H₈FNO₂
MDL Number:MFCD00143236
:181.1637 g/mol
UN:3439
ADR:6.1,III
:GHS06
Specification Test Results
- Appearance:Light yellow to Brown powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:141.0 to 145.0 deg-C
Specification Test Results
Frequently Bought Together