
2-(Methylthio)benzothiazole ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:615-22-5
:
Formula:C₈H₇NS₂
MDL Number:MFCD00005784
:181.2779 g/mol
Boiling Pt:177°C
EINECS:210-417-1
:GHS07
Specification Test Results
- Appearance:White to Light yellow powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:44.0 to 48.0 deg-C
- Solubility in Methanol:almost transparency
Specification Test Results
Frequently Bought Together