

1,2-Epoxycyclopentane ≥97.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:285-67-6
:
Formula:C₅H₈O
MDL Number:MFCD00005161
:84.12 g/mol
Boiling Pt:102°C
Flash Pt:10°C
EINECS:206-005-6
UN:1993
ADR:3,II
:GHS02
:GHS07
Specification Test Results
- Appearance:Colorless to Light orange to Yellow clear liquid
- Purity (GC):min. 97.0 %
Specification Test Results
Frequently Bought Together