

1,2-Epoxycyclooctane ≥97.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:286-62-4
:
Formula:C₈H₁₄O
MDL Number:MFCD00005164
:126.196 g/mol
Boiling Pt:75°C
EINECS:206-010-3
UN:1325
ADR:4.1,III
:GHS02
:GHS07
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 97.0 %
- Melting point:53.0 to 57.0 deg-C
- Solubility in Methanol:almost transparency
Specification Test Results
Frequently Bought Together