Order Entry
Italy
ContactUsLinkComponent
Chlorobis(4-fluorophenyl)phosphine ≥98%
Corrosive
Chlorobis(4-fluorophenyl)phosphine ≥98%
Catalog # H26953.03
CAS Number:  
Corrosive
Chlorobis(4-fluorophenyl)phosphine ≥98%
Catalog # H26953.03
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 g

Specifications

About this item

 :
ADR:8,II
 :256.62 g/mol
UN:3265
 :GHS05
Storage Temperature:Ambient
CAS Number:23039-97-6
Formula:C₁₂H₈ClF₂P
MDL Number:MFCD04972302