Order Entry
Italy
ContactUsLinkComponent
1-Hydroxy-4-nitro-9,10-anthraquinone ≥97.0% (by GC, titration analysis)
1-Hydroxy-4-nitro-9,10-anthraquinone ≥97.0% (by GC, titration analysis)
Supplier:  TCI
Certificates
undefined
1-Hydroxy-4-nitro-9,10-anthraquinone ≥97.0% (by GC, titration analysis)
Supplier:  TCI

Specification Test Results

  • Appearance:
    Light yellow to Brown to Dark green powder to crystal
  • Purity (GC):
    min. 97.0 %
  • Purity (Neutralization titration):
    min. 97.0 %
  • Solubility in hot DMF:
    almost transparency

Specification Test Results

Frequently Bought Together