
1,5-Diamino-4,8-dihydroxy-9,10-anthraquinone ≥95.0% (by HPLC, total nitrogen)
Supplier: TCI
Certificates
About this item
CAS Number:145-49-3
:
Formula:C₁₄H₁₀N₂O₄
MDL Number:MFCD00013722
EINECS:205-655-8
:GHS07
Specification Test Results
- Appearance:Gray to Dark purple to Black powder to crystal
- Purity (HPLC):min. 95.0 area%
- Purity (with Total Nitrogen):min. 95.0 %
Specification Test Results
Frequently Bought Together