Order Entry
Italy
ContactUsLinkComponent
 
1,4-Diamino-2,3-dichloro-9,10-anthraquinone ≥93.0% (by HPLC)
 :  TCI
 
undefined
1,4-Diamino-2,3-dichloro-9,10-anthraquinone ≥93.0% (by HPLC)
 :  TCI

 

  • Appearance:
    Gray to Dark purple to Black powder to crystal
  • Purity (HPLC):
    min. 93.0 area%
  • Purity (with Total Nitrogen):
    min. 90.0 %

 

Frequently Bought Together