Inserisci ordine
Italy
ContactUsLinkComponent
4,4'-Stilbenedicarboxylic acid ≥95.0% (by HPLC)
Irritant
4,4'-Stilbenedicarboxylic acid ≥95.0% (by HPLC)
Catalog # TCIAS0557-25G
Supplier:  TCI
CAS Number:  
undefined
Irritant
4,4'-Stilbenedicarboxylic acid ≥95.0% (by HPLC)
Catalog # TCIAS0557-25G
Supplier:  TCI
CAS Number:  

Specifications

  • Tipo di conf.:
    Bottiglia di vetro
  • Conf:
    25 g
  • Seq:
    3308

Specifications

Specification Test Results

  • Appearance:
    White to Orange to Green powder to crystal
  • Purity (HPLC):
    min. 95.0 area%
  • Purity (Neutralization back titration):
    min. 98.0 %

Specification Test Results

About this item

 :
 :268.27 g/mol
EINECS:202-838-4
 :GHS07
CAS Number:100-31-2
Formula:C₁₆H₁₂O₄
MDL Number:MFCD00013994