Order Entry
Italy
Orders LinkContactUsLinkComponent
Estriol

Estriol

   Sustainable Options Available
Supplier:   Cayman Chemical
Estriol
Supplier:  Cayman Chemical
   Sustainable Options Available
Certificates

4 Options Available Below

Product Details & Documents
  • CAS Number:50-27-1
  • Formula:C₁₈H₂₄O₃
  • MDL Number:MFCD00003691
  • EINECS:200-022-2
Specifications for Product Family

Specifications

Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle for solids
    100 mg
    Log in for availability
    Log in for availability
    1 * 100 mg
    1 * 100 mg - 25,20 €
    25,20 €
    1 * 100 mg
       25,20 €

    Specifications:
      
    More
    Specifications
    Cat. No.
    CAYM10006484-100
      
  • Glass bottle for solids
    500 mg
    Log in for availability
    Log in for availability
    1 * 500 mg
    1 * 500 mg - 91,20 €
    91,20 €
    1 * 500 mg
       91,20 €

    Specifications:
      
    More
    Specifications
    Cat. No.
    CAYM10006484-500
      
  • Glass bottle for solids
    1 g
    Log in for availability
    Log in for availability
    1 * 1 g
    1 * 1 g - 152,00 €
    152,00 €
    1 * 1 g
       152,00 €

    Specifications:
      
    More
    Specifications
    Cat. No.
    CAYM10006484-1
      
  • Glass bottle for solids
    5 g
    Log in for availability
    Log in for availability
    1 * 5 g
    1 * 5 g - 650,00 €
    650,00 €
    1 * 5 g
       650,00 €

    Specifications:
      
    More
    Specifications
    Cat. No.
    CAYM10006484-5
      
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.