Order Entry
Italy
ContactUsLinkComponent
Tricyclohexylphosphine ∼18% ∼0.60 mol/L in toluene (contains Tricyclohexylphosphine Oxide)
Flammable
Irritant
Health
Tricyclohexylphosphine ∼18% ∼0.60 mol/L in toluene (contains Tricyclohexylphosphine Oxide)
Catalog # TCIAT1165-25ML
Supplier:  TCI
CAS Number:  
undefined
Flammable
Irritant
Health
Tricyclohexylphosphine ∼18% ∼0.60 mol/L in toluene (contains Tricyclohexylphosphine Oxide)
Catalog # TCIAT1165-25ML
Supplier:  TCI
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    25 mL
  • Seq:
    2315

Specifications

Specification Test Results

  • Appearance:
    Colorless to Almost colorless clear liquid
  • Concentration (Iodometric back titration):
    18.0 to 20.0 w/w%

Specification Test Results

About this item

 :
 :280.43 g/mol
Flash Pt:42°C
EINECS:220-069-2
UN:1993
ADR:3,II
 :GHS02
 :GHS07
 :GHS08
CAS Number:2622-14-2
Formula:C₁₈H₃₃P
MDL Number:MFCD00003853