Order Entry
Italy
ContactUsLinkComponent
Gel filtration media, Sephadex™ G-15
Gel filtration media, Sephadex™ G-15
Catalog # 17-0020-01
Supplier:  Cytiva
CAS Number:  
undefined
Gel filtration media, Sephadex™ G-15
Catalog # 17-0020-01
Supplier:  Cytiva
CAS Number:  

Specifications

  • Particle Size:
    40 - 120 µm
  • Pk:
    100 g
  • Description:
    Sephadex™ G-15
  • pH range:
    2 - 13
  • Matrix:
    Cross-linked dextran
  • Fractionation range:
    <1,5 x 10³ dextrans, <1,5 x 10³ globular proteins
  • Storage temperature:
    4 - 30 °C
  • Type:
    Solid

Specifications

About this item

Sephadex™ G-15 is a well established gel filtration medium for desalting and buffer exchange of peptides and small biomolecules >1500 molecular weight.

  • Quickly desalts, removes contaminants and transfers to a new buffer in a single step
  • Suitable for buffer exchange and cleanup of biological samples, for example peptides, small proteins, and oligosaccharides larger than 1500 molecular weight
  • Efficient removal of contaminants such as salts, dyes, and radioactive labels

Sephadex™ is a gel filtration medium prepared by crosslinking dextran with epichlorohydrin. Different types of Sephadex™ differ in their degree of cross-linking and hence in their degree of swelling and their molecular fractionation range. Sephadex™ G-15 is one of six different G-types ranging from G-10 for small molecules to G-100 for very large biomolecules.