Size exclusion chromatography columns, PL aquagel-OH™ preparative
Supplier: VARIAN
Certificates
About this item
These PL aquagel-OH preparative columns are used in fractionation of wide variety of water-soluble samples based on their size in solution.
- High loading maximises sample throughput
- Up to 10 times scale-up maximises yield
These columns and associated guard columns enable rapid and convenient scale-up from analytical separations. The columns are packed with the same robust macroporous particles as the analytical column range.
Frequently Bought Together