Order Entry
Italy
Orders LinkContactUsLinkComponent
Spring tubing clips, SAMCO
Spring tubing clips, SAMCO
Supplier:   MURRAY S
Spring tubing clips, SAMCO
Supplier:  MURRAY S
Certificates
Spring tubing clips, SAMCO
2 Options Available Below
Product Details & Documents
These spring tubing clips are made from chemical and stain resistant acetal copolymer and stainless steel.
  • Operates with parallel action to provide optimum closure of rubber and flexible PVC tubing
  • Size 8 accepts tubing up to 8 mm outside diameter
  • Size 12 accepts tubing up to 12 mm outside diameter
Product Family Options
Product Information
  • Spanning width
    Availability
    Price
  • 8 mm
    Log in for availability
    Log in for availability
    1 * 100 pezzi
    1 * 100 pezzi - 368,00 €
    368,00 €
    1 * 100 pezzi
       368,00 €
  • 12 mm
    Log in for availability
    Log in for availability
    1 * 100 pezzi
    1 * 100 pezzi - 415,00 €
    415,00 €
    1 * 100 pezzi
       415,00 €