Ice & Meat Samplers
Sterilisable stainless steel samplers, that are mostly used in the food industry.
Meat Sampler
Meat Samplers for microbiological sampling of carcass surfaces.
Ice Sampler
Samplers for semi-solid and frozen substances. Made from V4A so autoclavable and sterilisable.