Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
Rs.0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
India
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
FLASK VOL CL-A 250 ML WM CLR CODE (RED) 1 * 1 items
Catalog # RANKFV-WMCC-0250R
Supplier:
RANKEM
New Product
Certificates
Expand image
FLASK VOL CL-A 250 ML WM CLR CODE (RED) 1 * 1 items
Catalog # RANKFV-WMCC-0250R
Supplier:
RANKEM
New Product
Certificates
Related Information
Documentation
coc_RANKFV-WMCC-0250R_RD5KWV101
coc_RANKFV-WMCC-0250R_RD5KWV102
coc_RANKFV-WMCC-0250R_RD5KWV103
coc_RANKFV-WMCC-0250R_RD5KWV104
coc_RANKFV-WMCC-0250R_RD5KWV105
More
Documentation
Frequently Bought Together
VWR®, Nitrile Gloves
Supplier: VWR Collection
VWR® Serological Pipette, Standard Line
Supplier: VWR Collection
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
VWR® Facial Tissues
Supplier: VWR Collection
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
VWR® NITRILE LIGHT, Nitrile Gloves
Supplier: VWR Collection
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection
Acetonitrile, anhydrous (max. 0.003% H₂O) ≥99.95%, HiPerSolv CHROMANORM® Reag. USP, Reag. Ph. Eur., ACS, super gradient grade suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
Nitrile gloves, Kimtech™ Purple Nitrile™
Supplier: KIMBERLY CLARK