Order Entry
India
ContactUsLinkComponent
384 square well, ps, 'flat' clear bottom, black plate, 100/cs free of detectable dnase/rnase well volume of 120 µl ansi/sbs standard dimensions fully traceable through a defined lot number system 1 * 100 items
Catalog # 732-4438
Supplier:  VWR Collection
New Product
384 square well, ps, 'flat' clear bottom, black plate, 100/cs free of detectable dnase/rnase well volume of 120 µl ansi/sbs standard dimensions fully traceable through a defined lot number system 1 * 100 items
Catalog # 732-4438
Supplier:  VWR Collection