Order Entry
India
ContactUsLinkComponent
RECIRCULATING CHILLER, BENCHTOP, 0.4 KW, -20 TO 30C, CENTRIFUGAL PUMP 24.6 LPM/1.14 BAR, COLOR TOUCHSCREEN, UV LIGHT FLUID CLEANING, LIQUID LEVEL ON DISPLAY, 5 LANGUAGES, SELF-DIAGNOSTIC TROUBLESHOOTING, 200-240V/50HZ 1 * 1 items
Catalog # 462-0649
Supplier:  VWR Collection
New Product
RECIRCULATING CHILLER, BENCHTOP, 0.4 KW, -20 TO 30C, CENTRIFUGAL PUMP 24.6 LPM/1.14 BAR, COLOR TOUCHSCREEN, UV LIGHT FLUID CLEANING, LIQUID LEVEL ON DISPLAY, 5 LANGUAGES, SELF-DIAGNOSTIC TROUBLESHOOTING, 200-240V/50HZ 1 * 1 items
Catalog # 462-0649
Supplier:  VWR Collection