Order Entry
India
ContactUsLinkComponent
The VWR® Nano Spectrophotometer LS is an economical nano-spectrophotometer with LED light source and silicon photocell detector. Wa velengths of 260 and 280 nm allow for quick absorption measurements of DNA, RNA and Protein sample drops. Includes tou 1 * 1 items
Catalog # 77610-710
Supplier:  VWR Collection
New Product
The VWR® Nano Spectrophotometer LS is an economical nano-spectrophotometer with LED light source and silicon photocell detector. Wa velengths of 260 and 280 nm allow for quick absorption measurements of DNA, RNA and Protein sample drops. Includes tou 1 * 1 items
Catalog # 77610-710
Supplier:  VWR Collection