Order Entry
India
ContactUsLinkComponent
The VWR Microplate Reader is designed for visible range absorbance readings using 96-well plates. It incorporates a 7 inch touch screen and is a stand-alone instrument which does not require a PC for operation. Data transfer can be done using a USB s 1 * 1 items
Catalog # VWRI210-000168
Supplier:  VWR Collection
New Product
The VWR Microplate Reader is designed for visible range absorbance readings using 96-well plates. It incorporates a 7 inch touch screen and is a stand-alone instrument which does not require a PC for operation. Data transfer can be done using a USB s 1 * 1 items
Catalog # VWRI210-000168
Supplier:  VWR Collection