Order Entry
India
ContactUsLinkComponent
VWR PP BIO-RED AUTOCLAVE BAG 34L, 1SIDE BLACK PRINT+AUTOCLAVE INDICATOR, SIZE:610X765MMX75µM, PACKING:50PCS/PACK,5PACKS/CARTON WITH TWIST TIES 1 * 250 items
Catalog # 129-4213
Supplier:  VWR Collection
New Product
VWR PP BIO-RED AUTOCLAVE BAG 34L, 1SIDE BLACK PRINT+AUTOCLAVE INDICATOR, SIZE:610X765MMX75µM, PACKING:50PCS/PACK,5PACKS/CARTON WITH TWIST TIES 1 * 250 items
Catalog # 129-4213
Supplier:  VWR Collection