Order Entry
India
ContactUsLinkComponent
FLASK VOLUMETRIC 25 ML AMBER COLOUR WITH ROFLUMILAST PRINTED ON BULB OF FLASK, GREEN COLOUR CODE ON NECK OF FLASK, SOCKET SIZE 10/19, TOL. 0.04 ML, CLASS A AS PER IS 915 1 * 1 items
Catalog # RANKFVA-WMCC-0025R
Supplier:  RANKEM
New Product
FLASK VOLUMETRIC 25 ML AMBER COLOUR WITH ROFLUMILAST PRINTED ON BULB OF FLASK, GREEN COLOUR CODE ON NECK OF FLASK, SOCKET SIZE 10/19, TOL. 0.04 ML, CLASS A AS PER IS 915 1 * 1 items
Catalog # RANKFVA-WMCC-0025R
Supplier:  RANKEM