Order Entry
India
ContactUsLinkComponent
FLASK VOLUMETRIC 10 ML AMBER COLOUR WITH ROFLUMILAST PRINTED ON BULB OF FLASK, SOCKET SIZE 10/19, TOL. ±0.04 ML WIDE NECK, CLASS A AS PER IS 915 1 * 1 items
Catalog # RANKFVA-WMCC-0010R
Supplier:  RANKEM
New Product
FLASK VOLUMETRIC 10 ML AMBER COLOUR WITH ROFLUMILAST PRINTED ON BULB OF FLASK, SOCKET SIZE 10/19, TOL. ±0.04 ML WIDE NECK, CLASS A AS PER IS 915 1 * 1 items
Catalog # RANKFVA-WMCC-0010R
Supplier:  RANKEM